Skip to Content
Merck
All Photos(4)

Key Documents

H0541

Sigma-Aldrich

Anti-HOXB7 (C-terminal) antibody produced in rabbit

~1.0 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-HHO.C1, Anti-HOX2, Anti-HOX2C, Anti-Hox-2.3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~24 kDa

species reactivity

human

concentration

~1.0 mg/mL

technique(s)

immunoprecipitation (IP): 2.5-5 μg using lysates of HEK-293T cells over expressing human HOXB7
indirect immunofluorescence: 1-2 μg/mL using using paraformaldehyde fixed HEK-293T cells over expressing human HOXB7
western blot: 1-2 μg/mL using extracts of HEK-293T cells over expressing human HOXB7

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... HOXB7(3217)
mouse ... Hoxb7(15415)

General description

Homeobox B7 (HOXB7) gene is mapped to human chromosome 17q21.32. In vertebrates, 39 HOX genes are organized into four clusters (HOXA-HOXD), located on different chromosomes (7p15, 17q21.2, 12q13, and 2q31). HOX proteins have a highly conserved 60-amino-acid region (the homeobox).

Specificity

Anti-HOXB7 (C-terminal) recognizes human HOXB7 (also known as HOX2; HOX2C; HHO.

Application

Anti-HOXB7 (C-terminal) antibody produced in rabbit may be used in immunoblotting.

Biochem/physiol Actions

The homeobox with DNA-binding activity, contributes to their activity as transcription factors. Hox genes are evolutionarily conserved transcription factors, which act to control important development pathways involved in the morphogenesis of the embryo. HOXB7 is expressed in various human hematopoietic cell lines and tissues as well as in proliferating melanocytes. In addition to roles in normal development, altered homeobox gene function or expression is implicated in carcinomas associated with breast, prostate, kidney, colon, skin and brain.

Physical form

Solution in 0.01 M phos­phate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

For continuous use, store at 2–8 °C for up to one month. For extended storage, freeze in working aliquots at –20 °C. Repeated freezing and thawing, or storage in “frost-free” freezers, is not recommended. If slight turbidity occurs upon prolonged storage, clarify solution by centrifugation before use. Working dilutions should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dong Gao et al.
American journal of physiology. Cell physiology, 315(5), C675-C686 (2018-08-02)
Metastatic cutaneous squamous cell carcinoma (CSCC) is a major cause of death associated with nonmelanoma skin cancer. The involvement of homeobox B7 ( HOXB7) in cancers has been reported. Thus, the current study intends to explore the effect of HOXB7
Elisabetta Manuela Foppiani et al.
Stem cell research & therapy, 10(1), 101-101 (2019-03-21)
The ex vivo expansion potential of mesenchymal stromal/stem cells (MSC) together with their differentiation and secretion properties makes these cells an attractive tool for transplantation and tissue engineering. Although the use of MSC is currently being tested in a growing
M T Lewis
Breast cancer research : BCR, 2(3), 158-169 (2001-03-16)
Both normal development and neoplastic progression involve cellular transitions from one physiological state to another. Whereas much is being discovered about signal transduction networks involved in regulating these transitions, little progress has been made in identifying the higher order genetic
Yang Yang et al.
Molecular medicine reports, 16(3), 2773-2778 (2017-07-06)
Osteosarcoma is the most common primary malignant bone cancer in children and adolescents. Unfortunately, treatment failures are common due to metastasis and chemore-sistance, however, the underlying molecular mechanism remains unclear. Accumulating evidence indicated that the homeobox B7 (HOXB7) gene was
Nobumoto Tomioka et al.
Cancer genetics and cytogenetics, 201(1), 6-14 (2010-07-17)
Unlike the case with some other solid tumors, whole genome array screening has not revealed prognostic genetic aberrations in primary gastric cancer. Comparative genomic hybridization (CGH) using bacterial artificial chromosome (BAC) arrays for 56 primary gastric cancers resulted in identification

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service