Skip to Content
Merck
All Photos(1)

Key Documents

GW21965A

Sigma-Aldrich

Anti-NDUFA10 (ab1) antibody produced in chicken

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-CI-42KD, Anti-NADH dehydrogenase

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
CHF 275.00

CHF 275.00


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
50 μG
CHF 275.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

CHF 275.00


Estimated to ship on29 May 2025


Request a Bulk Order

biological source

chicken

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

manufacturer/tradename

Genway 15-288-21965A

technique(s)

indirect ELISA: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NDUFA10(4705)

Immunogen

Immunogen Sequence: GI # 4758768, sequence 21-140
Recombinant NADH dehydrogenase (ubiquinone) 1 α subcomplex, 10, 42kDa

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service