Skip to Content
Merck
All Photos(1)

Key Documents

GW20088F

Sigma-Aldrich

Anti-HDL antibody produced in chicken

affinity isolated antibody, buffered aqueous solution

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
CHF 366.00

CHF 366.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 μG
CHF 366.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

CHF 366.00


Check Cart for Availability

Request a Bulk Order

biological source

chicken

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

manufacturer/tradename

Genway 15-288-20088F

technique(s)

western blot: suitable

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... HSD11B1(3290)

Immunogen

HDL human native protein.

Application

Anti-HDL antibody produced in chicken is suitable for western blotting analysis at a dilution of 1:500, for tissue or cell staining at a dilution of 1:200.

Biochem/physiol Actions

11β-hydroxysteroid dehydrogenase type 1 is an NADPH-dependent enzyme highly expressed in metabolic tissues that include liver, adipose tissue and the central nervous system. It may contribute to the loss of muscle strength with age and is a key component of sarcopenia. Inhibition of HDL gene may have some therapeutic potential in sarcopenia. HDL gene variation may affect the physiological cortisol levels and the severity of age-related osteoporosis. It is involved in regenerating active glucocorticoids, that are selectively elevated in adipose tissue in human obesity and metabolic syndrome, both associated with chronic low-grade inflammation. HDL acts as an oxidoreductase to yield active glucocorticoids, cortisol or corticosterone. It is expressed in normal human epidermal keratinocytes (NHEKs) and negatively regulates proliferation of NHEKs.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alixe H M Kilgour et al.
PloS one, 8(12), e84057-e84057 (2014-01-07)
Sarcopenia, the loss of muscle mass and function with age, is associated with increased morbidity and mortality. Current understanding of the underlying mechanisms is limited. Glucocorticoids (GC) in excess cause muscle weakness and atrophy. We hypothesized that GC may contribute
Jirong Wang et al.
European journal of cancer prevention : the official journal of the European Cancer Prevention Organisation (ECP), 22(6), 523-528 (2013-09-26)
Glucocorticoid hormones have been reported to contribute to the regulation of cellular proliferation and differentiation and to inhibit the growth of cells in several colon tumors and adenocarcinoma cell lines. As a regulator of glucocorticoid levels, type I isoform HSD11B1
Heide Siggelkow et al.
The Journal of clinical endocrinology and metabolism, 99(2), E293-E302 (2013-11-29)
Higher physiological cortisol levels may increase the risk of age-related osteoporosis. We hypothesized that common polymorphisms in the cortisol synthesis enzyme 11β-hydroxysteroid dehydrogenase (HSD11B) may cause interindividual variations in cortisol levels and age-related bone loss. We performed a retrospective study
Cristina L Esteves et al.
The Journal of clinical endocrinology and metabolism, 99(1), E160-E168 (2013-11-19)
Levels of 11β-hydroxysteroid dehydrogenase type 1 (11β-HSD1), which regenerates active glucocorticoids, are selectively elevated in adipose tissue in human obesity and metabolic syndrome, both conditions associated with chronic low-grade inflammation. 11β-HSD1 expression is induced by proinflammatory cytokines in a variety
Saori Itoi et al.
Biochemical and biophysical research communications, 440(2), 265-270 (2013-09-24)
The endogenous glucocorticoid, cortisol, is released from the adrenal gland in response to various stress stimuli. Extra-adrenal cortisol production has recently been reported to occur in various tissues. Skin is known to synthesize cortisol through a de novo pathway and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service