GERPN8511
Nucleon PhytoPure
Cytiva RPN8511, pack of 50 × 1.0 g
About This Item
Recommended Products
packaging
pack of 50 × 1.0 g
manufacturer/tradename
Cytiva RPN8511
Related Categories
General description
Application
Features and Benefits
- PhytoPure resin binds plant polysaccharides ensuring high yields of high-quality plant genomic DNA in only 1 h.
- Two kit sizes provide economical scale-up using the same phenol-free protocol.
Storage and Stability
Analysis Note
Storage Class Code
12 - Non Combustible Liquids
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service