Skip to Content
Merck
All Photos(1)

Documents

GERPN3685

ECF Substrate

Cytiva RPN3685, pack of 1 ea

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352204
NACRES:
NA.83

packaging

pack of 1 ea

manufacturer/tradename

Cytiva RPN3685

shipped in

wet ice

storage temp.

2-8°C

Related Categories

General description

AlkPhos Direct Labeling and Detection Systems are based on the rapid, direct labeling of DNA or RNA probes with thermostable alkaline phosphatase. CDP-Star or ECF detection can be used. Each system includes the following reagents, sufficient for 25 or 50 labeling reactions and detection of 2500 cm2 or 5000 cm2 membrane (depending on product ordered): labeling reagent, crosslinker, control DNA, reaction buffer, hybridization buffer, blocking agent, CDP-Star or ECF detection reagents.

Application

DNA or RNA probes are labeled during a 30 min reaction and resulting probes can be used without purification. Hybridization stringency can be controlled by temperature as well as salt concentration. Detection is rapid, taking only 30 min from completion of hybridization to application of detection reagent.

Features and Benefits

  • Probes are labelled during a 30 minute reaction, resulting in probes that can be used without purification.
  • Hybridization stringency can be controlled by temperature as well as salt concentration. Detection is rapid, taking only 30 minutes from completion of hybridization to application of detection reagent.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

ECF is a trademark of Cytiva

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Storage Class Code

12 - Non Combustible Liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service