Documents
GE28-9068-50
Hyperfilm MP
W × L 18 cm × 24 cm, pack of 50 ea (individual film envelopes)
About This Item
UNSPSC Code:
41105300
NACRES:
NB.22
sterility
non-sterile
packaging
pack of 50 ea (individual film envelopes)
manufacturer/tradename
Cytiva 28906850
W × L
18 cm × 24 cm
Related Categories
General description
  • Hyperfilm MP is a multipurpose film ideal for use in both blots and sequencing experiments.
  • Compatible with all commonly used radioisotopes.
  • Intended for radiometric or chemiluminescent applications.
  • Provides optimal clarity for reliable high-resolution image quality.
  • Easy to use.
Storage and Stability
Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Legal Information
Hyperfilm is a trademark of Cytiva
Storage Class Code