Skip to Content
Merck
All Photos(1)

Key Documents

GE17-0671-01

Sigma-Aldrich

Mono Q® PC 1.6/5

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
NACRES:
SB.52

ligand

-OCH2CH(OH)CH2OCH2CH(OH)CH2N+(CH3)3

description

Ion Exchanger Type (value)

packaging

pkg of 1 ea

parameter

725 psi

bed size

1.6 mm × 50 mm

bed volume

0.1 mL

column I.D.

1.6 mm

matrix

monodisperse porous polystyrene/divinyl benzene beads

matrix active group

ammonium, quaternary phase

avg. part. size

10 μm

cleaning

1-14

working range

2-12

separation technique

anion exchange

General description

Mono Q® PC 1.6/5 are precision columns (0.10 ml) prepacked with MonoBeads. The columns are designed for fast, sensitive, and high-resolution ion exchange separations of biomolecules at micropreparative scale.

Features and Benefits

  • Monodisperse 10 μm porous beads yield high resolution, dynamic capacity, reproducibility, and durability.
  • Maximum resolution is achieved from the efficiency of small, perfectly spherical monodispersed particles optimally packed in columns coupled with the excellent selectivities of the Q ion exchangers.
  • Compatible with ÄKTA® design, FPLC System, and other high-performance LC systems.
  • MonoBeads are also available as Mono P® for chromatofocusing

Storage and Stability

Store at 4 to 30 °C (20% Ethanol)

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

FPLC is a trademark of Cytiva
Mono P is a registered trademark of Cytiva
Mono Q is a registered trademark of Cytiva
ÄKTA is a registered trademark of Cytiva

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Storage Class Code

3 - Flammable liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service