Documents
GBLRD476684
Grace Bio-Labs wound splint, w/ suture sites
clear silicone, O.D. × I.D. × thickness 14 mm × 10 mm × 0.5 mm
Synonym(s):
silicone wound splints, suture site template
About This Item
UNSPSC Code:
41122600
NACRES:
NB.22
material
clear silicone
sterility
(sterile)
sterile
packaging
pkg of 20 ea (2 splints per pouch)
manufacturer/tradename
Grace Bio-Labs 476684
O.D. × I.D. × thickness
14 mm × 10 mm × 0.5 mm
General description
Grace Bio-Labs has worked with researchers to develop and manufacture wound splint devices that aid in the understanding of wound and tissue repair for animal research and the testing of new therapies for safety and efficacy.
Wound splints are constructed of silicone and include suture sites for increased precision in affixing on or within. These devices enable reproducible and reliable experimental conditions for wound healing in animal research.
The splints are offered in a variety of shapes and sizes (visible stitch holes / suture sites, 5 mm dia.), shipped sterile and ready to use.