Skip to Content
Merck
All Photos(1)

Documents

G9660

Supelco

Gelatin veronal buffer−EDTA

Synonym(s):

GVB-EDTA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

sterility

sterile-filtered

drug control

Home Office Schedule 3; regulated under CDSA - not available from Sigma-Aldrich Canada

storage temp.

2-8°C

Components

Contains 141 mM NaCl, 0.1% gelatin, 1.8 mM sodium barbital, 3.1 mM barbituric acid and 10 mM EDTA, pH 7.3-7.4.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service