Skip to Content
Merck
All Photos(1)

Documents

G9018

Sigma-Aldrich

Glass beads, unwashed

150-212 μm (70-100 U.S. sieve)

Synonym(s):

Glass beads

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41100000
NACRES:
NB.22

particle size

150-212 μm (70-100 U.S. sieve)

Related Categories

Application

Glass beads, unwashed have been used:
  • for lysing Clostridium difficile cells to perform assays of β-glucuronidase
  • in granular bed collection substrate which is used to connect downstream of the impactor in the nanoparticle respiratory deposition (NRD) sampler and provide a collection of particles
  • for the disintegration of leaf material to form fine powder for DNA extraction

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ada Ching et al.
BMC genetics, 3, 19-19 (2002-10-09)
Recent studies of ancestral maize populations indicate that linkage disequilibrium tends to dissipate rapidly, sometimes within 100 bp. We set out to examine the linkage disequilibrium and diversity in maize elite inbred lines, which have been subject to population bottlenecks

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service