Skip to Content
Merck
All Photos(2)

Key Documents

G7261

Sigma-Aldrich

L-Glutamic acid γ-(7-amido-4-methylcoumarin)

fluorescent amino acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H16N2O5
CAS Number:
Molecular Weight:
304.30
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

Assay

≥98% (TLC)

form

powder

technique(s)

HPLC: suitable

color

white to light yellow

storage temp.

−20°C

SMILES string

CC1=CC(=O)Oc2cc(NC(=O)CCC(N)C(O)=O)ccc12

InChI

1S/C15H16N2O5/c1-8-6-14(19)22-12-7-9(2-3-10(8)12)17-13(18)5-4-11(16)15(20)21/h2-3,6-7,11H,4-5,16H2,1H3,(H,17,18)(H,20,21)

InChI key

JPOAPPISZAGCAO-UHFFFAOYSA-N

Biochem/physiol Actions

L-Glutamic acid γ-(7-amido-4-methylcoumarin) (gammaGlu-AMC) may be useful as a fluorogenic substrate for the detection, differentiation or characterization of gamma-glutamyltransferase(s)/transpeptidase(s).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H D Shine et al.
Neurochemical research, 6(4), 453-463 (1981-04-01)
A new fluorometric assay was developed for the measurement of gamma-glutamyl transpeptidase (gamma-GTP). The assay utilizes as a substrate the synthetic compound 7-gamma-glutamylamido-4-methyl coumarin which is cleaved by gamma-GTP to yield the highly fluorescent product 7-amino-4-methyl coumarin. Optimal excitation and
K D Hammond et al.
Annals of clinical biochemistry, 22 ( Pt 4), 376-380 (1985-07-01)
Activities and isoenzyme patterns of gamma-glutamyl transferase (GGT) were studied in serum samples from fifteen Bantu males suffering from primary hepatoma; the results were compared with those obtained from normal samples. Enzyme activities were significantly higher in hepatoma patients than
L Sacchetti et al.
Clinical chemistry, 34(2), 419-422 (1988-02-01)
In this electrophoretic procedure for measuring isoenzymes of gamma-glutamyltransferase, patterns obtained are highly reproducible and the analytical imprecision (CV) ranges from 1.10% to 6.17%. A cellulose acetate support is used, at 220 V for 40 min. Sharply resolved isoenzyme bands
E Prusak et al.
Clinica chimica acta; international journal of clinical chemistry, 107(1-2), 21-26 (1980-10-23)
Using 7-(gamma-L-glutamyl)-4-methyl-coumarylamide as the fluorogenic substrate a simple and sensitive method for the assay of transferase activity of gamma-glutamyl-transferase in human blood serum and some other biological fluids is described. The substrate was synthesized with a good yield by the
A sensitive fluorimetric assay for gamma-glutamyl transferase.
G D Smith et al.
Analytical biochemistry, 100(1), 136-139 (1979-11-15)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service