All Photos(1)
About This Item
Linear Formula:
NH2C(=NH)NH2 · ½H2CO3
CAS Number:
Molecular Weight:
90.08
Beilstein:
3628359
EC Number:
MDL number:
UNSPSC Code:
12352204
Recommended Products
mp
>300 °C (lit.)
SMILES string
NC(N)=N.NC(N)=N.OC(O)=O
InChI
1S/2CH5N3.CH2O3/c3*2-1(3)4/h2*(H5,2,3,4);(H2,2,3,4)
InChI key
STIAPHVBRDNOAJ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
replaced by
Product No.
Description
Pricing
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Dam. 1
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service