Skip to Content
Merck
All Photos(1)

Key Documents

G1278

Sigma-Aldrich

Galantide

≥90% (HPLC)

Synonym(s):

M15

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C104H151N25O26S
CAS Number:
Molecular Weight:
2199.53
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.26

Assay

≥90% (HPLC)

form

powder

technique(s)

drug transporter assay: suitable

color

white

UniProt accession no.

storage temp.

−20°C

SMILES string

CSCC[C@H](NC(=O)[C@H](CC(C)C)NC(=O)CNC(=O)[C@H](Cc1ccccc1)NC(=O)[C@H](Cc2ccccc2)NC(=O)[C@H](CCC(N)=O)NC(=O)[C@H](CCC(N)=O)NC(=O)[C@@H]3CCCN3C(=O)CNC(=O)[C@H](CC(C)C)NC(=O)[C@H](CC(C)C)NC(=O)[C@H](Cc4ccc(O)cc4)NC(=O)CNC(=O)[C@H](C)NC(=O)[C@H](CO)NC(=O)[C@H](CC(N)=O)NC(=O)[C@H](CC(C)C)NC(=O)[C@@H](NC(=O)[C@H](Cc5c[nH]c6ccccc56)NC(=O)CN)[C@@H](C)O)C(N)=O

InChI

1S/C104H151N25O26S/c1-54(2)39-70(91(142)113-52-87(139)129-37-20-27-80(129)103(154)120-69(33-35-82(107)134)93(144)119-68(32-34-81(106)133)94(145)124-76(44-61-23-16-13-17-24-61)99(150)123-74(43-60-21-14-12-15-22-60)92(143)112-51-86(138)116-71(40-55(3)4)95(146)118-67(89(109)140)36-38-156-11)121-96(147)72(41-56(5)6)122-98(149)75(45-62-28-30-64(132)31-29-62)117-85(137)50-111-90(141)58(9)114-102(153)79(53-130)127-100(151)78(47-83(108)135)125-97(148)73(42-57(7)8)126-104(155)88(59(10)131)128-101(152)77(115-84(136)48-105)46-63-49-110-66-26-19-18-25-65(63)66/h12-19,21-26,28-31,49,54-59,67-80,88,110,130-132H,20,27,32-48,50-53,105H2,1-11H3,(H2,106,133)(H2,107,134)(H2,108,135)(H2,109,140)(H,111,141)(H,112,143)(H,113,142)(H,114,153)(H,115,136)(H,116,138)(H,117,137)(H,118,146)(H,119,144)(H,120,154)(H,121,147)(H,122,149)(H,123,150)(H,124,145)(H,125,148)(H,126,155)(H,127,151)(H,128,152)/t58-,59+,67-,68-,69-,70-,71-,72-,73-,74-,75-,76-,77-,78-,79-,80-,88-/m0/s1

InChI key

TZOJVPDIYKRJSM-GKPUQKAJSA-N

Gene Information

human ... GAL(51083)

Amino Acid Sequence

Gly-Trp-Thr-Leu-Asn-Ser-Ala-Gly-Tyr-Leu-Leu-Gly-Pro-Gln-Gln-Phe-Phe-Gly-Leu-Met-NH2

Biochem/physiol Actions

Galanin, a neuropeptide, mediates its physiological effects through three G protein-coupled receptor (GCPR) subtypes, GalR1, GalR2, and GalR3. Galantide, C7, M35, and M40 are Galanin receptor antagonists used to differentiate and characterize Galanin receptor signaling pathways that impact processes such as acute pancreatitis.
Galanin receptor antagonist

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Charis E Teh et al.
Nature communications, 7, 13353-13353 (2016-11-20)
The linear ubiquitin chain assembly complex (LUBAC) is essential for innate immunity in mice and humans, yet its role in adaptive immunity is unclear. Here we show that the LUBAC components HOIP, HOIL-1 and SHARPIN have essential roles in late
Shang-Mian Yie et al.
British journal of cancer, 115(12), 1555-1564 (2016-11-23)
The production of autoantibodies against tumour-associated antigens (TAAs) is believed to reflect greater immunologic reactivity in cancer patients and enhanced immune surveillance for cancer cells. Over the past few decades, a number of different TAAs and their corresponding autoantibodies have
Naresh Chandra et al.
Viruses, 11(5) (2019-05-01)
Human adenoviruses (HAdV) are the most common cause of ocular infections. Species B human adenovirus type 3 (HAdV-B3) causes pharyngoconjunctival fever (PCF), whereas HAdV-D8, -D37, and -D64 cause epidemic keratoconjunctivitis (EKC). Recently, HAdV-D53, -D54, and -D56 emerged as new EKC-causing
Estefania Contreras et al.
Nature communications, 10(1), 2869-2869 (2019-06-30)
Clostridial neurotoxins, including tetanus and botulinum neurotoxins, generally target vertebrates. We show here that this family of toxins has a much broader host spectrum, by identifying PMP1, a clostridial-like neurotoxin that selectively targets anopheline mosquitoes. Isolation of PMP1 from Paraclostridium
Danielle Moinier et al.
Frontiers in microbiology, 8, 1277-1277 (2017-07-28)
The chemical attack of ore by ferric iron and/or sulfuric acid releases valuable metals. The products of these reactions are recycled by iron and sulfur oxidizing microorganisms. These acidophilic chemolithotrophic prokaryotes, among which

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service