Skip to Content
Merck
All Photos(1)

Key Documents

F9175

Sigma-Aldrich

Flt-3/Flk-2 Ligand from mouse

≥97% (SDS-PAGE), recombinant, expressed in NSO cells, lyophilized powder, suitable for cell culture

Sign Into View Organizational & Contract Pricing

Select a Size

5 μG
CHF 282.00

CHF 282.00


Estimated to ship on23 May 2025


Request a Bulk Order

Select a Size

Change View
5 μG
CHF 282.00

About This Item

MDL number:
UNSPSC Code:
51111800
NACRES:
NA.32

CHF 282.00


Estimated to ship on23 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

recombinant

expressed in NSO cells

Assay

≥97% (SDS-PAGE)

form

lyophilized powder

potency

1.0-4.0 ng/mL

mol wt

~18 kDa

packaging

pkg of 5 μg

storage condition

avoid repeated freeze/thaw cycles

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

UniProt accession no.

storage temp.

−20°C

Gene Information

mouse ... Flt3l(14256)

General description

Flt3l (FMS-like tyrosine kinase 3 ligand) is a cytokine and a hematopoietic growth factor. It acts as a ligand for FLT3 receptor.[1] Alternative splicing of FLT3L results in both membrane-bound and soluble forms. Soluble form is a result of proteolytic cleavage of the membrane isoform. It has a wide range of tissue expression.[2]
Mouse and human Flt-3 ligand share approximately 72% homology and both demonstrate cross-species activity.

Biochem/physiol Actions

Flt3l (FMS-like tyrosine kinase 3 ligand) acts as a stimulant of hematopoietic progenitor cell proliferation. It facilitates the commitment of lymphoid lineage, and aids in dendritic cell (DC) development. It is involved in myeloblast differentiation, and drives skeletal muscle regeneration. It acts on p120RasGAP and suppresses Erk signaling, which in turn promotes myogenesis.[1] In lymphoid tissues, it modulates DC homeostasis, which is facilitated by steady state as well as proliferative T-cells.[2]

Quality

Heterogenous glycosylation causes recombinant mouse Flt-3 ligand to migrate as multiple bands from 18-32 kDa.

Physical form

Lyophilized from a 0.2 μm filtered solution in 50% acetonitrile and 0.1% trifluoroacetic acid containing 50 μg bovine serum albumin per 1 μg of cytokine.

Analysis Note

Bioactivity is measured by the ability to stimulate the proliferation of a Flt-3 transfected pro-B cell line.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S D Lyman et al.
Current opinion in hematology, 2(3), 177-181 (1995-05-01)
The flt3 ligand is a member of a small family of growth factors that stimulate the proliferation of hematopoietic cells. Other members of this family include Steel factor (also known as mast cell growth factor, stem cell factor, and kit
Yejing Ge et al.
Journal of cell science, 126(Pt 15), 3370-3379 (2013-05-25)
Various cues initiate multiple signaling pathways to regulate the highly coordinated process of skeletal myogenesis. Myoblast differentiation comprises a series of ordered events starting with cell cycle withdrawal and ending with myocyte fusion, with each step probably controlled by multiple
Yasuyuki Saito et al.
European journal of immunology, 43(6), 1651-1658 (2013-03-23)
Lymphoid-tissue dendritic cells (DCs) are short-lived and need to be continuously replenished from bone marrow-derived DC progenitor cells. Fms-related tyrosine kinase 3 is expressed during cellular development from hematopoietic progenitors to lymphoid-tissue DCs. Fms-related tyrosine kinase 3 ligand (Flt3L) is

Articles

Read article on hematopoietic cytokines and hematopoiesis

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service