Skip to Content
Merck
All Photos(1)

Key Documents

F1503

Sigma-Aldrich

Fructosazine

Synonym(s):

2,5-bis(D-Arabinotetrahydroxybutyl)pyrazine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H20N2O8
CAS Number:
Molecular Weight:
320.30
UNSPSC Code:
12352200

storage temp.

−20°C

InChI

1S/C12H20N2O8/c15-3-7(17)11(21)9(19)5-1-13-6(2-14-5)10(20)12(22)8(18)4-16/h1-2,7-12,15-22H,3-4H2/t7-,8-,9-,10-,11-,12-/m1/s1

InChI key

NPWQIVOYGNUVEB-PAUJSFGCSA-N

Looking for similar products? Visit Product Comparison Guide


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tomoko Shimamura et al.
Bioscience, biotechnology, and biochemistry, 67(2), 295-299 (2003-05-06)
XTT (3'-[1-[(phenylamino)-carbonyl]-3,4-tetrazolium]-bis(methoxy-6-nitro)benzenesulfonic acid hydrate) was reduced by incubated glucosamine hydrochloride. The XTT reducibility by incubated glucosamine was linearly related with the DNA-breaking activity. In order to elucidate the reaction mechanism, the glucosamine derivatives formed during the incubation process were separated
K Sumoto et al.
Chemical & pharmaceutical bulletin, 39(3), 792-794 (1991-03-01)
Two pyrazine derivatives [fructosazine (3) and deoxyfructosazine (6)] were simultaneously formed in a solution of D-glucosamine hydrochloride under various conditions. They showed deoxyribonucleic acid (DNA) strand breakage activity in plasmid pBR322 comparable to that of D-glucosamine. The DNA strand breakage
Nathaly Henry et al.
Talanta, 99, 816-823 (2012-09-13)
Fructosazine and 2,5-deoxyfructosazine are two natural chemicals with various applications as flavoring agents in food and tobacco industry; the 2,5-deoxyfructosazine has also anti-diabetic and anti-inflammatory activities. In order to quantify these compounds in natural samples such as plant or food
G Candiano et al.
Carbohydrate research, 184, 67-75 (1988-12-31)
The reaction of 2-amino-2-deoxy-D-glucose with lysine in water under simulated physiological conditions gave several browning products, with characteristic optical (lambda max 340 nm) and fluorescent properties (emission at 430 nm for excitation at 362 nm). The major product was isolated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service