Skip to Content
Merck
All Photos(1)

Documents

F0804

Sigma-Aldrich

Fetal Bovine Serum, Research Grade

non-USA origin, sterile-filtered, suitable for cell culture

Synonym(s):

Fetal Bovine Serum, Fetal Bovine Sera, Fetal Calf Sera, FBS, FCS, sera, serum

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352207
NACRES:
NA.71

biological source

bovine fetus

sterility

sterile-filtered

product line

FBS Classic

composition

hemoglobin (tested)

origin

non-USA origin

availability

available only in EU

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested
mycoplasma, tested
virus (BVD, IBR, PI3), tested negative

shipped in

dry ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Fetal bovine serum (FBS), a popular supplement, is mainly sourced from unborn calves. It contains nutrients, vitamins, hormones, transport proteins, and attachment, spreading, and growth factors.

Application

Fetal Bovine Serum, Research Grade has been used as a supplement:
  • of endothelial cell medium (ECMed) for culturing human umbilical vein endothelial cells (HUVEC)
  • in Dulbecco′s modified Eagle′s medium (DMEM) to resuspend human adipose tissue-derived stromal cells (ASC)
  • in the tissue culture medium 199 (TCM199) for in vitro oocyte maturation
  • in minimum essential medium (MEM) Earle′s medium for culturing Madin-Darby canine kidney (MDCK) II cells

Biochem/physiol Actions

Fetal bovine serum (FBS) is mainly used in organ, cell, and tissue culture. FBS is the first choice to be used as a supplement in all eukaryotic cell culture media. It supports the proliferation and viability of most cell types.

Analysis Note

  • Endotoxin and hemoglobin tested
  • Tested for the presence of bacteria, virus, and mycoplasma
  • Triple filtered with 0.1 micron membrane under aseptic conditions

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kayleigh M van Megen et al.
Frontiers in immunology, 10, 694-694 (2019-04-20)
Mesenchymal stromal cells (MSCs) are inherently immunomodulatory through production of inhibiting soluble factors and expression of immunosuppressive cell surface markers. We tested whether activated MSCs qualify for the induction of antigen-specific immune regulation. Bone marrow derived human MSCs were activated
Sara Kaliman et al.
Frontiers in physiology, 7, 551-551 (2016-12-10)
It is well accepted that cells in the tissue can be regarded as tiles tessellating space. A number of approaches were developed to find an appropriate mathematical description of such cell tiling. A particularly useful approach is the so called
J van der Valk et al.
Toxicology in vitro : an international journal published in association with BIBRA, 18(1), 1-12 (2003-11-25)
Fetal bovine serum (FBS) is a common supplement to in vitro culture media. A workshop was organized to discuss whether or not fetuses might suffer when blood is withdrawn, and to discuss serum replacement methods. When bovine fetuses are exposed
Jan van der Valk
Science (New York, N.Y.), 375(6577), 143-144 (2022-01-14)
Ethical and possible reproducibility issues arise when using fetal bovine serum in cell culture media.
Simona Ranallo et al.
Nature communications, 8, 15150-15150 (2017-05-10)
A wide range of molecular devices with nanoscale dimensions have been recently designed to perform a variety of functions in response to specific molecular inputs. Only limited examples, however, utilize antibodies as regulatory inputs. In response to this, here we

Articles

Review the basics of using fetal bovine (or calf) serum for cell culture and how sourcing and regulations affect the price of sera for cell-based assays.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service