EP5910711000
Adapter for Eppendorf® S-4x400 Rotor
for 12x75 mm round-bottom tubes (FACS), pkg of 2 ea
Select a Size
CHF 310.00
Estimated to ship on02 May 2025
Select a Size
About This Item
CHF 310.00
Estimated to ship on02 May 2025
feature
Holds 12 x 75 mm tubes
for 12x75 mm round-bottom tubes (FACS)
packaging
pkg of 2 ea
manufacturer/tradename
Eppendorf® 5910711000
technique(s)
centrifugation: suitable
compatibility
for use with 400 mL round bucket
Looking for similar products? Visit Product Comparison Guide
General description
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service