EP5820768002
Adapter for Eppendorf® FA-45-12-17/FA-45-16-17/FA-45-20-17 Rotors
capacity, 1 x 1.5 - 2.0 mL vessels, pack of 4
All Photos(1)
About This Item
Recommended Products
feature
capacity 1 x 1.5 - 2.0 mL vessels
Holds 1.5/2.0 mL tubes
packaging
pack of 4
manufacturer/tradename
Eppendorf® 5820768002 (Global)
technique(s)
centrifugation: suitable
compatibility
for use with FA-45-16-17
for use with FA-45-20-17
for use with Rotor FA-45-12-17
Related Categories
Legal Information
Eppendorf is a registered trademark of Eppendorf AG