EP5810755000
Eppendorf® Adapter for 250 mL Rectangular Bucket
for use with 9 x 15 mL Falcon tubes, set of 2 pieces
Select a Size
CHF 217.00
Select a Size
About This Item
CHF 217.00
Recommended Products
feature
for use with 9 x 15 mL Falcon tubes
packaging
set of 2 pieces
manufacturer/tradename
Eppendorf® 5810755000
technique(s)
centrifugation: suitable
compatibility
for use with 250 mL rectangular buckets
Looking for similar products? Visit Product Comparison Guide
General description
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service