Skip to Content
Merck
All Photos(1)

Documents

EP5427754008

Rotor for Eppendorf® 5430/5430R Centrifuges

FA-45-48-11, aerosol tight, to hold, 48 x 1.5/2.0 mL tubes, lid

Synonym(s):

Lid for Eppendorf® FA-45-48-11 Rotor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103909
NACRES:
NB.82

feature

lid
to hold 48 x 1.5/2.0 mL tubes
aerosol tight
fixed angle rotor

packaging

pack of 1 ea

manufacturer/tradename

Eppendorf® 5427754008 (Global)

parameter

12,700 rpm
18,210 × g max. RCF

technique(s)

centrifugation: suitable

compatibility

for use with centrifuge models 5430/5430R

Looking for similar products? Visit Product Comparison Guide

General description

  • Fixed angle rotor holds 48 x 1.5/2.0 mL tubes.
  • Features a QuickLock rotor lid for ergonomic lid locking.
  • The aerosol-tight lock facilitates safe centrifugation of hazardous samples.
  • Compatible with the 5430 and 5430R with serial numbers at or above 16,000.

Legal Information

Eppendorf is a registered trademark of Eppendorf AG

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service