Key Documents
EP5424706002
Eppendorf® Rotor F-45-18-11-Kit
incl.lid
Select a Size
1 EA
CHF 641.00
CHF 641.00
Check Cart for Availability
Select a Size
Change View
1 EA
CHF 641.00
About This Item
UNSPSC Code:
41103909
NACRES:
NB.82
CHF 641.00
Check Cart for Availability
feature
to hold 18 x 1.5/2.0 mL tubes
to hold 18 x spin columns
incl.lid
packaging
pack of 1 ea
manufacturer/tradename
Eppendorf® 5424706002
parameter
15,000 max. RCF (18,111 xg)
technique(s)
centrifugation: suitable
compatibility
for use with centrifuge models 5424/5424R
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)