Key Documents
EP4920610000
Eppendorf® Reference 2 lower part
single-channel, lower part with separate serial number, violet, size 0.5-5 μL
Select a Size
1 EA
CHF 67.50
CHF 67.50
Check Cart for Availability
Select a Size
Change View
1 EA
CHF 67.50
About This Item
UNSPSC Code:
41121500
NACRES:
NB.01
CHF 67.50
Check Cart for Availability
feature
single-channel
lower part with separate serial number
packaging
pkg of 1 ea
manufacturer/tradename
Eppendorf® 4920610000
size
0.5-5 μL
color
violet
General description
The name, Reference, stands for extraordinary precision and accuracy, a long service lifes and an ergonomic design. The new Reference 2 boasts these proven premium characteristics with its state-of-the-art technolgy, making it a reliable partner for you and your work. The time saving single-button operation is unrivalled in the new multichannel versions.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)