EP022639188
ADAPTER FOR EPPENDORF(R) A-4-38 ROTOR
(holds 1 X 15 ML CONICAL TUBE, FOR 100 ML ROUND BUCKET), pkg of 2 ea
About This Item
Recommended Products
feature
(holds 1 X 15 ML CONICAL TUBE, FOR 100 ML ROUND BUCKET)
packaging
pack of 2 ea
pkg of 2 ea
manufacturer/tradename
Eppendorf® 022639188 (North America)
technique(s)
centrifugation: suitable
compatibility
for use with 100 mL round bucket
Related Categories
General description
- Round bucket design
- Designed for 15mL Falcon centrifuge tubes
- Use with Eppendorf rotor model A-4-38
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service