EP0030711122
Eppendorf® Cell Culture Flask T-75
polystyrene flask, working volume 20 mL, Tissue Culture (TC)-treated surface, sterile, with cap, filtered, case of 80 ea (16 bags × 5 flasks)
About This Item
Recommended Products
material
canted neck
polystyrene flask
screw top plastic cap
sterility
sterile
feature
DNase free
RNase free
endotoxin free
vented
with cap filtered
packaging
case of 80 ea (16 bags × 5 flasks)
manufacturer/tradename
Eppendorf® 0030711122
W × D × H
163.0 cm × 82.0 mm × 46.6 mm
maximum volume
279.8 mL
surface area
77.9 cm2
working volume
20 mL
binding type
Tissue Culture (TC)-treated surface
Looking for similar products? Visit Product Comparison Guide
General description
Cell accessibility
A unique ConvexAccess neck geometry significantly facilitates the access to the growth area and makes cell seeding and media exchange more convenient, safe and reliable.
Improved contamination protection
The new hydrophobic volume filter pad is more efficient than standard filter membranes
Corrugated cap
Anti–rolling cap with corrugation and clear position marks in plug–seal caps to prevent undesired full–closing
Features and Benefits
Optimized gas exchange and protection against contamination through high efficiency air filter technology
Defined arrested position on plug-seal caps to prevent undesired closing
Anti-rolling cap with corrugation facilitates cultivation steps when the cap needs to be set aside
100 % in-line control for leakage-free flasks and maximum safety
Direct surface identification on flask and cap
Sterile, pyrogen-, DNase-, RNase-, human and bacterial DNA-free. Non-cytotoxic
Can be centrifuged with the suitable adapters; refer to the manufacturer′s instruction.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service