Key Documents
EP0030626303
Eppendorf® Lobind Microplate 384/V-PP, 384 wells
sterile, pkg of 240 ea, white polypropylene, V-bottom clear wells
Select a Size
240 EA
CHF 1’810.00
CHF 1’810.00
Check Cart for Availability
Select a Size
Change View
240 EA
CHF 1’810.00
About This Item
UNSPSC Code:
41122107
NACRES:
NB.15
material:
V-bottom clear wells
white polypropylene
size:
384 wells
sterility:
sterile
CHF 1’810.00
Check Cart for Availability
material
V-bottom clear wells
white polypropylene
sterility
sterile
packaging
pkg of 240 ea
manufacturer/tradename
Eppendorf® 0030626303
technique(s)
PCR: suitable
size
384 wells
General description
Maximize your sample recovery with Eppendorf LoBind Plates. Eppendorf LoBind Plates are designed with a special two-component polymer mix that creates a hydrophilic surface to ensure optimal sample recovery rates. These plates are specifically designed for research or sensitive testing methods where they enable excellent results due to less sample binding to the plate surface.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)