Key Documents
EP0030132530
Eppendorf® Twin tec real-time PCR plates
blue plate, conical bottom white wells, semi-skirted, pkg of 25 ea
Select a Size
25 EA
CHF 211.00
CHF 211.00
Check Cart for Availability
Select a Size
Change View
25 EA
CHF 211.00
About This Item
UNSPSC Code:
41106300
NACRES:
NB.15
material:
blue plate
conical bottom white wells
size:
96 well
feature:
skirt (Semi skirted)
CHF 211.00
Check Cart for Availability
material
blue plate
conical bottom white wells
feature
skirt (Semi skirted)
packaging
pkg of 25 ea
manufacturer/tradename
Eppendorf® 0030132530
technique(s)
PCR: suitable
size
96 well
working volume
250 μL
General description
The limiting factor in low-volume real-time PCR (qPCR) is often the remaining fluorescence intensity. Eppendorf twin.tec real-time PCR plates are specially manufactured for qPCR, as they offer all the advantages of regular twin.tec plates, but with the added advantage of white wells. These reflect fluorescence much better than clear or frosted wells, so lower levels of fluorescence are still detectable with the same instrument – just by using the right consumables. The qPCR plates′ white wells also significantly reduce interfering background fluorescence and lead to increased consistency between replicates, and giving highly reproducible results. The wells are made from polypropylene – which guarantees fast heat transfer for fast assays – and the frame is made of rigid polycarbonate, to ensure easy and reliable handling – both in manual and automated assays.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)