EP0030131517
Eppendorf® plate lid
PCR clean, pkg of 80 ea
Select a Size
CHF 151.00
Estimated to ship on27 May 2025
Select a Size
About This Item
CHF 151.00
Estimated to ship on27 May 2025
Recommended Products
feature
PCR clean
packaging
pkg of 80 ea
manufacturer/tradename
Eppendorf® 003013517
Features and Benefits
- Sealing Mats are reusables and can be autoclave. Sealing mats are suitable for short-term storage
- Heat Sealing Film and Foil for continuous locking give the best evaporation protection
- Foil products are made of aluminum, can be pierced, and protect light-sensitive samples
- Film products are transparent and provide protection against unwanted punctures
- Eppendorf Plate Lid: stable, flexible protection of samples during short-term storage
- Storage Film and Foil glued seals for simple and dependable sealing during sample storage
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)