Key Documents
EP0030129628
Eppendorf twin.tec® PCR Plates 384
skirted, Forensic DNA Grade, working volume 40 μL, colorless, pack of 10 (individually wrapped)
Select a Size
10 EA
CHF 82.50
CHF 82.50
Check Cart for Availability
Select a Size
Change View
10 EA
CHF 82.50
About This Item
UNSPSC Code:
41106300
NACRES:
NB.15
material:
colorless
colorless wells
size:
384 well
feature:
skirt
Forensic DNA Grade
skirted
CHF 82.50
Check Cart for Availability
material
colorless
colorless wells
well size
384
feature
skirt
Forensic DNA Grade
skirted
packaging
pack of 10 (individually wrapped)
manufacturer/tradename
Eppendorf® 0030129628 (Global)
technique(s)
PCR: suitable
size
384 well
working volume
40 μL
General description
The Eppendorf twin.tec PCR plate combines extremely thin-walled polypropylene wells for optimal heat transfer to the sample, with robust polycarbonate frames for high rigidity. With raised well rims, twin.tec plates enable highly effective sealing to reduce the risk of cross-contamination.
Features and Benefits
  • Optimized rim for barcodes and automated work processes
  • Stackable
  • Within SBS footprint recommendations
  • Ideal for most common 384-well thermocyclers, particularly for Eppendorf Mastercycler X50 and Mastercycler nexus
  • 40 µL max. well volume A
  • Ideal for quantitative real-time PCR as well – OptiTrack matrix for faster sample identification and fewer pipetting errors
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
twin.tec is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)