Skip to Content
Merck
All Photos(1)

Key Documents

EP0030128869

Eppendorf® Twin tec PCR plates, 96 wells

colorless plate, conical bottom colorless wells, semi-skirted, pkg of 300 ea

Sign Into View Organizational & Contract Pricing

Select a Size

300 EA
CHF 1’770.00

CHF 1’770.00


Estimated to ship on27 April 2025



Select a Size

Change View
300 EA
CHF 1’770.00

About This Item

UNSPSC Code:
41106300
NACRES:
NB.15
material:
colorless plate
conical bottom colorless wells
maximum volume:
-250 μL
size:
96 wells
working volume:
-200 μL
sterility:
non-sterile
binding type:
low binding surface
feature:
skirt (Semi skirted)
skirt (semi-skirted)

CHF 1’770.00


Estimated to ship on27 April 2025


material

colorless plate
conical bottom colorless wells

sterility

non-sterile

feature

skirt (Semi skirted)
skirt (semi-skirted)

packaging

pkg of 300 ea

manufacturer/tradename

Eppendorf® 0030128869

technique(s)

PCR: suitable

maximum volume

-250 μL

size

96 wells

working volume

-200 μL

suitability

suitable for PCR
suitable for molecular biology

binding type

low binding surface

Looking for similar products? Visit Product Comparison Guide

General description

Choose Eppendorf twin.tec PCR plates for confidence in every PCR assay. twin.tec plates offer purity, rigidity, color, and other simple, smart features can make your daily life a lot easier. The rigid polycarbonate frame works well in automation and high-throughput processes, and the different colors mean you can quickly organize the different steps of your workflow.
The “Eppendorf” Twin tec PCR plates are designed for single-use PCR applications. The plates are semi-skirted and equipped with thin-walled polypropylene wells and polycarbonate frames. These features offer durability, rigidity, and aid in automation and simplified handling during manual workflows.

Application

“Eppendorf” Twin tec PCR plates are suitable for use in:
  • Standard PCR
  • Real-time PCR
  • High Throughput PCR
  • Small Volume Sample Handling

Features and Benefits

  • The “Eppendorf” Twin tec PCR plates are semi-skirted, allowing for easy labeling and barcoding.
  • They are compatible with most thermal cyclers.
  • Each well when used with cap strips has a maximum well volume of 250 µL, providing ample capacity.
  • They offer improved well-to-well tolerance.
  • The raised well rims contribute to effective sealing, while simultaneously reducing the potential for cross-contamination.
  • These plates are certified to be free of prevalent PCR contaminants such as DNA, DNAse, RNAse, and PCR inhibitors.

Legal Information

Eppendorf is a registered trademark of Eppendorf AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service