Skip to Content
Merck
All Photos(1)

Key Documents

EP0030128770

Eppendorf® Twin tec PCR plates, 96 wells

colorless plate, conical bottom colorless wells, skirted, pkg of 300 ea

Synonym(s):

96-Well PCR Plates, PCR Plates, Twin tec PCR Plates

Sign Into View Organizational & Contract Pricing

Select a Size

300 EA
CHF 1’770.00

CHF 1’770.00


Check Cart for Availability


Select a Size

Change View
300 EA
CHF 1’770.00

About This Item

UNSPSC Code:
41106300
NACRES:
NB.15
material:
colorless plate
conical bottom colorless wells
size:
96 wells
sterility:
non-sterile
feature:
skirt

CHF 1’770.00


Check Cart for Availability

material

colorless plate
conical bottom colorless wells

sterility

non-sterile

feature

skirt

packaging

pkg of 300 ea

manufacturer/tradename

Eppendorf® 0030128770

technique(s)

PCR: suitable

size

96 wells

well maximum volume

150 μm

Looking for similar products? Visit Product Comparison Guide

General description

Choose Eppendorf twin.tec PCR plates for confidence in every PCR assay. twin.tec plates offer purity, rigidity, color, and other simple, smart features can make your daily life a lot easier. The rigid polycarbonate frame works well in automation and high-throughput processes, and the different colors mean you can quickly organize the different steps of your workflow.
Eppendorf® twin.tec PCR plates provide confidence in every polymerase chain reaction (PCR) assay by ensuring purity, rigidity, color, and other simple, smart features making your daily workflow a lot easier. Eppendorf Twin tec PCR plates, 96 wells are suitable for high-throughput applications and automated workflows.

Features and Benefits

  • The extremely thin-walled polypropylene wells ensure optimal heat transfer to the sample.
  • Robust polycarbonate frames offer high rigidity.
  • Raised rims enable highly effective sealing to reduce the risk of cross-contamination.
  • All polymerase chain reaction (PCR) consumables are made of high-purity plastics and are ‘PCR clean’. They are batch-tested and certified to be free of a range of common PCR contaminants, including DNA, DNAse, RNAse, and PCR inhibitors.
  • The plate′s edge is optimized for barcodes and automated work processes.
  • Meets SBS footprint recommendations (127.76 × 85.47mm ± 0.25mm) stackable
  • Eight notches in the plate skirt facilitate easy insertion and removal from the thermoblock, streamlining experimental workflows.
  • The plate′s specific design enables efficient PCR, even with low sample volumes. Maximum well volume 150 µL (when using cap strips)
  • Barcode functionality is available upon request for skirted and semi-skirted plates, enhancing sample identification.

Legal Information

Eppendorf is a registered trademark of Eppendorf AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service