Key Documents
EP0030120973
Eppendorf® micropestles
pkg of 10 ea
Select a Size
10 EA
CHF 54.10
CHF 54.10
Check Cart for Availability
Select a Size
Change View
10 EA
CHF 54.10
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
CHF 54.10
Check Cart for Availability
packaging
pkg of 10 ea
manufacturer/tradename
Eppendorf® 0030120973
General description
  • Eppendorf® micropestles are accessories for sample handling.
  • They are used for resuspending pellets and comprise of two sides for optimal fit in 1.5 mL and 2.0 mL vessels.
  • Autoclavable (121 °C, 20 min).
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)