Key Documents
EP0030077598
Eppendorf® ep Dualfilter T.I.P.S.®
PCR clean, capacity 1-10 mL, sterile, pack of 100 ea (individually wrapped)
Select a Size
100 EA
CHF 98.50
CHF 98.50
Check Cart for Availability
Select a Size
Change View
100 EA
CHF 98.50
About This Item
UNSPSC Code:
41121600
NACRES:
NB.02
volume:
1—10 mL
sterility:
sterile
feature:
DNase free
RNase free
filter
packaging:
pack of 100 ea (individually wrapped)
manufacturer/tradename:
Eppendorf® 0030077598
CHF 98.50
Check Cart for Availability
material
turquoise
sterility
sterile
feature
DNase free
RNase free
filter
packaging
pack of 100 ea (individually wrapped)
manufacturer/tradename
Eppendorf® 0030077598
technique(s)
PCR: suitable
capacity
1-10 mL
maximum volume
10 mL
volume
1—10 mL
General description
  • Eppendorf® ep Dualfilter T.I.P.S.® of 1-10 mL capacity comprise of two filter layers that provide dual protection against aerosols and biomolecules to guard your pipette and sample.
  • They are well-suited for applications with samples that contain detergents.
  • It is available as PCR clean/Sterile (sterile and pyrogen-free), followed by continuous inspection of each production lot by an independent laboratory which guarantees high purity and quality.
  • Ultrahomogenous surface for maximum reproducibility.
  • Ultrahydrophobic surface for minimal sample loss.
  • Significantly reduced foam formation during pipetting.
  • Extreme resistance to chemicals.
  • Available in PCR clean and Eppendorf Quality.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
ep Dualfilter T.I.P.S. is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)