Key Documents
EP0030010027
Eppendorf® epT.I.P.S.® G Singles, Biopur®
volume (0.2-20 μL), L (40 mm), medium gray Tray, pkg of 100 tips, individually wrapped
Select a Size
100 EA
CHF 37.80
CHF 37.80
Check Cart for Availability
Select a Size
Change View
100 EA
CHF 37.80
About This Item
UNSPSC Code:
41121600
NACRES:
NB.02
volume:
(0.2-20 μL)
sterility:
sterile; γ-irradiated
feature:
DNase free
individually wrapped
packaging:
pack of 100 ea (individually wrapped)
pkg of 100 tips
manufacturer/tradename:
Eppendorf® 0030010027
CHF 37.80
Check Cart for Availability
material
colorless
sterility
sterile; γ-irradiated
feature
DNase free
individually wrapped
packaging
pack of 100 ea (individually wrapped)
pkg of 100 tips
manufacturer/tradename
Eppendorf® 0030010027
L
(40 mm)
maximum volume
20 μL
volume
(0.2-20 μL)
color
medium gray Tray
binding type
non-treated surface
Legal Information
Biopur is a registered trademark of Eppendorf AG
Eppendorf is a registered trademark of Eppendorf AG
epT.I.P.S. is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)