Skip to Content
Merck
All Photos(1)

Key Documents

EMU216481

Sigma-Aldrich

MISSION® esiRNA

targeting mouse Vcan

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GTCATAGCAAGCCCAGAGCAGCTGTTTGCCGCCTATGAGGATGGATTTGAGCAGTGTGATGCAGGATGGCTGTCTGATCAAACTGTCAGATATCCCATACGGGCTCCCCGAGAGGGCTGTTACGGAGACATGATGGGGAAGGAAGGGGTTCGGACCTATGGATTCCGCTCTCCCCAGGAAACCTATGATGTGTATTGTTATGTGGATCATCTGGATGGCGATGTGTTCCACATCACTGCTCCCAGTAAGTTCACCTTCGAGGAGGCCGAAGCAGAGTGTACAAGCAGGGATGCGAGGCTGGCGACTGTTGGAGAACTTCAGGCAGCTTGGAGAAATGGCTTTGACCAATGCGATTACGGCTGGCTGTCGGATGCCAGCGTGCGGCACCCTGTGACTGTGGCCAGGGCCCAGTGTGGAGGAGGTCTACTTGGGGTGAGAACCCTGTATCGTTTTGAGAACCAGACATGCTTCCCTCTCCCTGATAGCAGATTTGATGCCTACTGCTTTAAA

Ensembl | mouse accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

Related Categories

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zi Wang et al.
Oncology reports, 33(6), 2981-2991 (2015-04-16)
The systematic application of antiangiogenic therapy remains an issue of concern, mainly due to the hypoxic and inflammatory changes in the tumor microenvironment elicited by antiangiogenic therapy. Versican, a 'bridge' connecting inflammation with tumor progression as well as playing a
Lu-lu Xu et al.
Respiratory physiology & neurobiology, 215, 58-63 (2015-05-23)
COPD lung is characterized by loss of alveolar elastic fibers and an increase in the chondroitin sulfate (CS) matrix proteoglycan versican V1 (V1). V1 is a known inhibitor of elastic fiber deposition and this study investigates the effects of knockdown
Jon M Carthy et al.
Cardiovascular pathology : the official journal of the Society for Cardiovascular Pathology, 24(6), 368-374 (2015-09-24)
Versican is a versatile and highly interactive chondroitin sulfate proteoglycan that is found in the extracellular matrix (ECM) of many tissues and is a major component of developing and developed lesions in atherosclerotic vascular disease. In this paper, we present
Naoko Arichi et al.
Oncoscience, 2(2), 193-204 (2015-04-11)
In the current study, we investigated a combination of docetaxel and thalidomide (DT therapy) in castration-resistant prostate cancer (CRPC) patients. We identified marker genes that predict the effect of DT therapy. Using an androgen-insensitive PC3 cell line, we established a
Pamela A Havre et al.
BMC cancer, 13, 517-517 (2013-11-05)
CD26/dipeptidyl peptidase IV (DPPIV) is a multifunctional membrane protein with a key role in T-cell biology and also serves as a marker of aggressive cancers, including T-cell malignancies. Versican expression was measured by real-time RT-PCR and Western blots. Gene silencing

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service