Skip to Content
Merck
All Photos(1)

Documents

EMU044441

Sigma-Aldrich

MISSION® esiRNA

targeting mouse Ppargc1a

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

AACAAGCACTTCGGTCATCCCTGTCAAGCTGTGTTTGACGACAAATCAGACAAGACCAGTGAACTAAGGGATGGCGACTTCAGTAATGAACAATTCTCCAAACTACCTGTGTTTATAAATTCAGGACTAGCCATGGATGGCCTATTTGATGACAGTGAAGATGAAAGTGATAAACTGAGCTACCCTTGGGATGGCACGCAGCCCTATTCATTGTTCGATGTGTCGCCTTCTTGCTCTTCCTTTAACTCTCCGTGTCGAGACTCAGTGTCACCACCGAAATCCTTATTTTCTCAAAGACCCCAAAGGATGCGCTCTCGTTCAAGATCCTTTTCTCGACACAGGTCGTGTTCCCGATCACCATATTCCAGGTCAAGATCAAGGTCCCCAGGCAGTAGATCCTCTTCAAGATCCTGTTACTACTATGAATCAAGCCACTACAGACACCGCACACACCGCAATTCTCCCTTGTATGTGAGATCACGTTCAAGGTCACCCTACAGCCGTAGGCCCAGGTACGACAGCTATGAAGCCTATGAGCACGAAAGGCTCAAGAGGGATGAATACCGC

Ensembl | mouse accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Biochem/physiol Actions

Ppargc1a (peroxisome proliferator-activated receptor γ coactivator 1-α) is a transcriptional co-activator. It is down-regulated in prostate cancer and is linked with disease progression. It plays an important role in mitochondrial function and biogenesis. Changes in the methylation pattern of the gene promoter causes changes in the mitochondrial DNA content. It controls endothelial homeostasis by participating in endothelial nitric oxide (NO) synthase (eNOS) activity and NO production. It plays an important role in insulin signaling. It is responsible for the activation of gluconeogenesis in hepatocytes, regulating thermogenesis in brown adipose tissue, fatty acid oxidation in the heart. It also controls reactive oxygen species production.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

PGC-1a ameliorates AngiotensinII-induced eNOS dysfunction in human aortic endothelial cells.
Li J
Vascular Pharmacology, 83, 90-90 (2016)
Epigenetic regulation of an adverse metabolic phenotype in polycystic ovary syndrome: the impact of the leukocyte methylation of PPARGC1A promoter.
Zhao H
Fertility and Sterility, 107, 467-467 (2017)
The metabolic co-regulator PGC1a suppresses prostate cancer metastasis.
Torrano V, et. al.
Nature Cell Biology, 18, 645-645 (2016)
Lack of direct evidence for natural selection at the candidate thrifty gene locus, PPARGC1A.
Cadzow M
BMC Medical Genetics, 17, 80-80 (2016)
Suppression of endothelial PGC-1a is associated with hypoxia-induced endothelial dysfunction and provides a new therapeutic target in pulmonary arterial hypertension.
Ye JX
American Journal of Physiology. Lung Cellular and Molecular Physiology, 310, L1233-L1233 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service