Skip to Content
Merck
All Photos(1)

Key Documents

EMU021521

Sigma-Aldrich

MISSION® esiRNA

targeting mouse Tiam1

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
CHF 249.00
50 μG
CHF 443.00

CHF 249.00


Check Cart for Availability


Select a Size

Change View
20 μG
CHF 249.00
50 μG
CHF 443.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

CHF 249.00


Check Cart for Availability

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CAGATGGCAAGAGGGAGAAGGAAGTGGTCTTACCCAGTGTCCACCAGCACAACCCCGACTGTGACATTTGGGTCCATGAATATTTCACTCCATCCTGGTTCTGTCTACCCAACAACCAGCCAGCCTTGACGGTTGTCCGGCCAGGGGACACTGCGAGGGACACCTTGGAGCTCATTTGCAAGACACATCAACTGGATCATTCCGCCCATTACCTGCGCCTGAAATTCCTAATGGAGAACAGAGTGCAGTTCTACATCCCGCAGCCCGAGGAGGACATTTACGAGCTGCTTTACAAAGAAATTGAAATCTGTCCAAAAGTCACCCAGAATATCCACATTGAGAAGTCAGACGCGGCCGCTGATAATTACGGGTTTTTGCTTTCTTCTGTGGATGAAGATGGCATTCGAAGGCTCTACGTGAACAGTGTCAAGGAAACCGGGTTAG

Ensembl | mouse accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Minjuan Wu et al.
Clinical science (London, England : 1979), 129(7), 575-588 (2015-05-23)
The homing ability and secretory function of mesenchymal stem cells (MSCs) are key factors that influence cell involvement in wound repair. These factors are controlled by multilayer regulatory circuitry, including adhesion molecules, core transcription factors (TFs) and certain other regulators.
Mina Ding et al.
OncoTargets and therapy, 11, 4367-4375 (2018-08-14)
T-cell lymphoma invasion and metastasis inducing factor 1 (Tiam1) is known to be involved in tumor progression. However, its molecular roles and mechanism in pancreatic ductal adenocarcinoma (PDAC) remain unclear. The purpose of this study is to determine Tiam1 expression
G Zhu et al.
Oncogene, 34(49), 5971-5982 (2015-03-10)
Epidermal growth factor receptor (EGFR) signaling regulates cell growth and survival, and its overactivation drives cancer development. One important branch of EGFR signaling is through activation of GTPase Rac1, which further promotes cell proliferation, survival and cancer metastasis. Here, we
Helen J Whalley et al.
Nature communications, 6, 7437-7437 (2015-06-17)
Centrosome separation is critical for bipolar spindle formation and the accurate segregation of chromosomes during mammalian cell mitosis. Kinesin-5 (Eg5) is a microtubule motor essential for centrosome separation, and Tiam1 and its substrate Rac antagonize Eg5-dependent centrosome separation in early

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service