Skip to Content
Merck
All Photos(1)

Documents

EMU005581

Sigma-Aldrich

MISSION® esiRNA

targeting mouse Timp1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CGAATCAACGAGACCACCTTATACCAGCGTTATAAGATCAAGATGATGACTAAGATGCTAAAAGGATTCAAGGCTGTGGGAAATGCCGCAGATATCCGGTACGCCTACACCCCAGTCATGGAAAGCCTCTGTGGATATGCCCACAAGTCCCAGAACCGCAGTGAAGAGTTTCTCATCACGGGCCGCCTAAGGAACGGGAAATTTCACATCAATGCCTGCAGCTTCTTGGTTCCCTGGCGTACTCTGAGCCCTGCTCAGCAAAGAGTTTTCTCAAAAAAGAACTATAGTGCTGGCTGTGGGGTGTGCACAGTGTTTCCCTGTTTATCTATCCCTTGCAAACTGGAGAGTGACACTCACTGTTTGTGGACGGATCAGGTCCTCGTGGGCTCTGAGGACTACCAGAGCCGTC

Ensembl | mouse accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

mouse ... Timp1(21857)

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiujuan Yao et al.
Respirology (Carlton, Vic.), 20(5), 730-738 (2015-05-02)
Interleukin (IL)-25 has been implicated in the pathogenesis of human asthma by inducing a Th2 cytokine response, but its possible role in the development of airway remodelling is less clear. We developed a murine surrogate of chronic airway inflammation induced
Rosemarie Chirco D'Angelo et al.
Molecular cancer research : MCR, 12(9), 1324-1333 (2014-06-05)
Tissue inhibitor of metalloproteinase-1 (TIMP-1) regulates intracellular signaling networks for inhibition of apoptosis. Tetraspanin (CD63), a cell surface binding partner for TIMP-1, was previously shown to regulate integrin-mediated survival pathways in the human breast epithelial cell line MCF10A. In the
Robert Ramer et al.
Biochemical pharmacology, 91(2), 202-216 (2014-07-01)
Cannabinoids inhibit tumor neovascularization as part of their tumorregressive action. However, the underlying mechanism is still under debate. In the present study the impact of cannabinoids on potential tumor-to-endothelial cell communication conferring anti-angiogenesis was studied. Cellular behavior of human umbilical

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service