Skip to Content
Merck
All Photos(1)

Key Documents

EHU115141

Sigma-Aldrich

MISSION® esiRNA

targeting human HSPA8

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
CHF 249.00
50 μG
CHF 443.00

CHF 249.00


Check Cart for Availability


Select a Size

Change View
20 μG
CHF 249.00
50 μG
CHF 443.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

CHF 249.00


Check Cart for Availability

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

AACCGAACCACTCCAAGCTATGTCGCCTTTACGGACACTGAACGGTTGATCGGTGATGCCGCAAAGAATCAAGTTGCAATGAACCCCACCAACACAGTTTTTGATGCCAAACGTCTGATTGGACGCAGATTTGATGATGCTGTTGTCCAGTCTGATATGAAACATTGGCCCTTTATGGTGGTGAATGATGCTGGCAGGCCCAAGGTCCAAGTAGAATACAAGGGAGAGACCAAAAGCTTCTATCCAGAGGAGGTGTCTTCTATGGTTCTGACAAAGATGAAGGAAATTGCAGAAGCCTACCTTGGGAAGACTGTTACCAATGCTGTGGTCACAGTGCCAGCTTACTTTAATGACTCTCAGCGTCAGGCTACCAAAGATGCTGGAACTATTGCTGGTCTCAATGTACTTAGAATTATTAATGAGCCAACTGCTGCTG

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Midori Ikezaki et al.
Biochemical and biophysical research communications, 527(2), 481-488 (2020-04-28)
Heat-shock cognate protein 70 (Hsc70), a molecular chaperone, is involved in multiple cellular functions. We previously demonstrated that Hsc70 is required for TGF-β-induced Smad signaling in mesenchymal-like NRK-49F cells. In the present study, to compare the Hsc70 functions in TGF-β-related
Guan Sun et al.
Neuromolecular medicine, 21(1), 33-41 (2019-01-05)
Heat shock cognate protein 70 (Hsc70) is a key mediator for the maintenance of intracellular proteins and regulates cellular activities. And it is elevated in various tumor tissues including glioma, which is closely related to the malignancy and poor prognosis
Guan Sun et al.
Journal of cellular biochemistry, 120(6), 10707-10714 (2019-03-01)
Migration and invasion are often recognized as the main reasons for the high recurrence and death rates of glioma and limit the efficacy of surgery and other antitumor therapies. In this study, we found over activation of heat shock cognate
Ximeng Yang et al.
Scientific reports, 8(1), 11707-11707 (2018-08-05)
We previously found diosgenin, an herbal drug-derived steroid sapogenin, to be remarkably effective at restoring Aβ-induced axonal degeneration and improving memory function in model of Alzheimer's disease (AD), 5XFAD mouse. In this study, we investigated the downstream signaling of diosgenin
Nerea Allende-Vega et al.
Scientific reports, 9(1), 5637-5637 (2019-04-06)
Eliminating mutant p53 (mt p53) protein could be a useful strategy to treat mt p53 tumors and potentially improve the prognosis of cancer patients. In this study, we unveil different mechanisms that eliminate p53-R248Q, one of the most frequent mutants

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service