Skip to Content
Merck
All Photos(1)

Key Documents

EHU095161

Sigma-Aldrich

MISSION® esiRNA

targeting human NCAM1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GTGTGGTTACAGGCGAGGATGGCAGTGAGTCAGAGGCCACCGTCAACGTGAAGATCTTTCAGAAGCTCATGTTCAAGAATGCGCCAACCCCACAGGAGTTCCGGGAGGGGGAAGATGCCGTGATTGTGTGTGATGTGGTCAGCTCCCTCCCACCAACCATCATCTGGAAACACAAAGGCCGAGATGTCATCCTGAAAAAAGATGTCCGATTCATAGTCCTGTCCAACAACTACCTGCAGATCCGGGGCATCAAGAAAACAGATGAGGGCACTTATCGCTGTGAGGGCAGAATCCTGGCACGGGGGGAGATCAACTTCAAGGACATTCAGGTCATTGTGAATGTGCC

Ensembl | human accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yongguang Yang et al.
BMC cell biology, 11, 78-78 (2010-10-20)
The proliferation and final density of Sertoli cells in the testis are regulated by hormones and local factors. Glial cell line-derived neurotrophic factor (GDNF), a distantly related member of the transforming growth factor-β superfamily, and its receptor subunits GDNF family
Chunlei Yu et al.
Toxicology mechanisms and methods, 26(9), 635-643 (2016-11-08)
Curcuma phaeocaulis Val. is a Chinese medicinal herb that is contraindicated during pregnancy for over a thousand years in China. The aims of the present study were to evaluate the effect of curcumol (one of the major components of C.
Natacha Coppieters et al.
Brain research, 1710, 199-208 (2018-12-26)
The neural cell adhesion molecule (NCAM) is a transmembrane protein involved in major cellular processes. The addition of polysialic acid (PSA), a post-translational modification (PTM) almost exclusively carried by NCAM, alters NCAM properties and functions and is therefore tightly regulated.
Mizuki Sumida et al.
The Journal of biological chemistry, 290(21), 13202-13214 (2015-03-10)
As acidic glycocalyx on primary mouse microglial cells and a mouse microglial cell line Ra2, expression of polysialic acid (polySia/PSA), a polymer of the sialic acid Neu5Ac (N-acetylneuraminic acid), was demonstrated. PolySia is known to modulate cell adhesion, migration, and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service