Skip to Content
Merck
All Photos(1)

Key Documents

EHU086641

Sigma-Aldrich

MISSION® esiRNA

targeting human SLC12A9

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
CHF 249.00
50 μG
CHF 443.00

CHF 249.00


Estimated to ship on20 April 2025



Select a Size

Change View
20 μG
CHF 249.00
50 μG
CHF 443.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

CHF 249.00


Estimated to ship on20 April 2025


description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CTGAACACACTGGCTGCTGTGGTCACTGTCTTCTACCTGGTGGCCTATGCTGCCGTGGACCTGTCCTGCCTGAGCCTGGAGTGGGCCTCGGCCCCCAACTTCCGCCCCACCTTCAGCCTGTTCTCCTGGCACACCTGCCTGCTGGGGGTGGCCTCCTGCCTGCTCATGATGTTCCTCATCAGTCCTGGCGCGGCTGGTGGCTCCCTGCTCCTCATGGGTCTGCTGGCTGCCCTGCTCACCGCGCGAGGAGGCCCCAGTAGCTGGGGCTATGTCAGCCAGGCCTTGCTTTTCCACCAGGTGCGTAAGTATCTGCTTCGGCTGGACGTCCGGAAGGATCACGTGAAGTTCTGGCGGCCCCAGCTGCTGCTCCTGGTGGGGAACCCCCGGGGCGCCCTGCCTCTGCTGCGGTTGGCCAACCAGCTTAAGAAGGGG

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anna E Vilgelm et al.
EBioMedicine, 24, 43-55 (2017-10-17)
Antagonists of MDM2-p53 interaction are emerging anti-cancer drugs utilized in clinical trials for malignancies that rarely mutate p53, including melanoma. We discovered that MDM2-p53 antagonists protect DNA from drug-induced damage in melanoma cells and patient-derived xenografts. Among the tested DNA
Desheng Zhong et al.
Journal of cellular biochemistry, 115(9), 1624-1635 (2014-05-03)
Pan-Bcl-2 family inhibitor obatoclax has been demonstrated to be effective against various cancers, of which the mechanism of action is not fully understood. In this study, we demonstrate that obatoclax suppressed esophageal cancer cell viability with concomitant G1/G0-phase cell cycle
S Kagawa et al.
Oncogene, 34(18), 2347-2359 (2014-06-17)
Notch activity regulates tumor biology in a context-dependent and complex manner. Notch may act as an oncogene or a tumor-suppressor gene even within the same tumor type. Recently, Notch signaling has been implicated in cellular senescence. Yet, it remains unclear

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service