Skip to Content
Merck
All Photos(1)

Key Documents

EHU083211

Sigma-Aldrich

MISSION® esiRNA

targeting human TLE1

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
CHF 249.00
50 μG
CHF 443.00

CHF 249.00


Estimated to ship on02 June 2025



Select a Size

Change View
20 μG
CHF 249.00
50 μG
CHF 443.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

CHF 249.00


Estimated to ship on02 June 2025


description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

TCCTCGTCAGTGCTTAGCTGTGACATCTCTGTGGATGATAAGTACATAGTCACTGGCTCGGGGGACAAGAAGGCTACAGTCTATGAAGTCATCTACTGAAAACATTATGTGGTTTAACGTTTATAGTTGAATTGGGCCAAAATGTTTCGAATTTATAGAAATAGAAAAGTTGTAACTTTAAAAGAGAAAAAAAATTACAAACACCTGTTTCCAAACCTTGACAGAAAACTACTTTGAGTCTACAAAGAGGAGGCGACAAGTCCATCAGCAGAAAGTCACCTGTCTACATAGACCAAATGGAGCACCAAGGCCAAGCGGACAGAGGGGCCATGGGTTGTAGGATTGAGGAACGGAATCTGCCGACTCACATGACAGCCCATTCTTTCTTTCTGGGTGATCTGGG

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wei Chen et al.
Bioscience, biotechnology, and biochemistry, 84(6), 1176-1182 (2020-03-03)
Liver damage induced by ischemia/reperfusion (I/R) remains a primary issue in multiple hepatic surgeries. Innate immune-mediated inflammatory responses during the reperfusion stage aggravate the injury. Nevertheless, the detailed mechanism of hepatic I/R has not been fully clarified yet. Our research
Federica De Paoli et al.
FEBS letters, 590(1), 43-52 (2016-01-15)
Macrophages display heterogeneous phenotypes, including the classical M1 proinflammatory and the alternative M2 anti-inflammatory polarization states. The transducin-like enhancer of split-1 (TLE1) is a transcriptional corepressor whose functions in macrophages have not been studied yet. We report that TLE1 is
Xin Yao et al.
Oncotarget, 8(42), 72235-72249 (2017-10-27)
The Transducin-like enhancer of split 1 (TLE1) corepressor protein is overexpressed in human lung tumors and is a putative lung-specific oncogene. However, the molecular mechanism underlying its oncogenic function remains to be delineated. Here, we report an important role of
Xin Yao et al.
PloS one, 9(7), e101564-e101564 (2014-07-09)
The mitochondrial Bit1 (Bcl-2 inhibitor of transcription 1) protein is a part of an apoptotic pathway that is uniquely regulated by integrin-mediated attachment. As an anoikis effector, Bit1 is released into the cytoplasm following loss of cell attachment and induces

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service