Skip to Content
Merck
All Photos(1)

Key Documents

EHU045151

Sigma-Aldrich

MISSION® esiRNA

targeting human C5

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
CHF 249.00
50 μG
CHF 443.00

CHF 249.00


Estimated to ship on22 April 2025



Select a Size

Change View
20 μG
CHF 249.00
50 μG
CHF 443.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

CHF 249.00


Estimated to ship on22 April 2025


description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

AAGCCAGCTCCGTGCTAATATCTCTCATAAAGACATGCAATTGGGAAGGCTACACATGAAGACCCTGTTACCAGTAAGCAAGCCAGAAATTCGGAGTTATTTTCCAGAAAGCTGGTTGTGGGAAGTTCATCTTGTTCCCAGAAGAAAACAGTTGCAGTTTGCCCTACCTGATTCTCTAACCACCTGGGAAATTCAAGGCGTTGGCATTTCAAACACTGGTATATGTGTTGCTGATACTGTCAAGGCAAAGGTGTTCAAAGATGTCTTCCTGGAAATGAATATACCATATTCTGTTGTACGAGGAGAACAGATCCAATTGAAAGGAACTGTTTACAACTATAGGACTTCTGGGATGCAGTTCTGTGTTAAAATGTCTGCTGTGGAGGGAATCTGCACTTCGGAAAG

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

human ... C5(727) , C5(727)

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gaurav Mehta et al.
Journal of immunology (Baltimore, Md. : 1950), 194(11), 5446-5454 (2015-04-29)
Rheumatoid arthritis (RA) is an inflammatory autoimmune joint disease in which the complement system plays an important role. Of the several components of complement, current evidence points to C5 as the most important inducer of inflammation. Several groups generated Abs
Xiaofei Yan et al.
Molecular and cellular biochemistry, 398(1-2), 95-104 (2014-09-14)
Excessive reactive oxygen species (ROS) generation has been implicated as one of main agents in ouabain-induced anticancer effect. Unfortunately, the signaling pathways under it are not very clarified. In the present study, we investigated the molecular mechanism involved in ouabain-induced
Annalisa Natalicchio et al.
Diabetologia, 58(6), 1260-1271 (2015-03-27)
The role of the redox adaptor protein p66(Shc) as a potential mediator of saturated fatty acid (FA)-induced beta cell death was investigated. The effects of the FA palmitate on p66(Shc) expression were evaluated in human and murine islets and in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service