Skip to Content
Merck
All Photos(1)

Key Documents

EHU044751

Sigma-Aldrich

MISSION® esiRNA

targeting human MACC1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CAAGGTGATTTCAAAGGAGCAAGTAATGTTTATGTCAGATAGTGTCTTTACAACCAGAAATCTTCTTGAACAGATTGTCCTGCCTTTAAAAAAATTGACTTATATCTACTCAGTTGTATTAACCTTGGTGTCAGAAAAAGTTTATGATTGGAAAGTTTTAGCTGATGTCCTGGGTTACTCACATCTGTCCCTGGAAGATTTTGATCAAATTCAAGCAGACAAAGAATCAGAGAAAGTTTCTTATGTTATAAAGAAGTTAAAGGAAGATTGCCACACAGAGAGAAATACAAGGAAGTTTCTGTATGAACTTATTGTGGCTCTTCTGAAAATGGATTGCCAAGAGTTAGTCGCACGTCTCATCCAAGAAGCTGCTGTTCTGACTTCAGCTGTCAAGCTTGGAAAAGG

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C J Wei et al.
Neoplasma, 67(3), 537-546 (2020-02-18)
Gastric cardia adenocarcinoma (GCA) is one of the most common types of cancer and the incidence is increasing globally. MicroRNAs (miRNAs) have been reported to play critical roles in the progression of GCA. However, the exact role of miR-638 in
Qiang Zhang et al.
Acta biochimica et biophysica Sinica, 50(8), 748-756 (2018-07-03)
One of the major obstacles hindering the treatment of lung cancer (LC) is chemoresistance; however, its mechanism remains unclear. The overexpression of the metastasis-associated in colon cancer 1 (MACC1) gene has been demonstrated to reverse chemoresistance. In the current study
Mingliang Lu et al.
Experimental and therapeutic medicine, 17(4), 2807-2814 (2019-03-25)
The mortality and incidence rates of colorectal cancer (CRC) vary widely worldwide. miR-338-3p inhibits tumor cell proliferation in several types of cancer, however, the role of miR-338-3p on CRC remains unknown. The aim of the current study was to investigate
Yaoqing Li et al.
Molecular medicine reports, 12(1), 426-434 (2015-03-05)
Metastasis-associated in colon cancer-1 (MACC1) is a newly identified gene that is involved in the development and progression of hepatocellular carcinoma (HCC), however its investigation has not been comprehensive. In the present study, in vitro techniques, including immunohistochemistry, western blotting
Aiko Sueta et al.
International journal of oncology, 46(5), 2143-2153 (2015-03-05)
The newly identified gene, metastasis‑associated in colon cancer 1 (MACC1), is suggested to be a transcriptional regulator of c‑Met, leading to cancer progression in colorectal cancer. To date however, little is known of the role of MACC1 in breast cancer.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service