Skip to Content
Merck
All Photos(1)

Key Documents

EHU007471

Sigma-Aldrich

MISSION® esiRNA

targeting human ST6GALNAC5

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
CHF 249.00
50 μG
CHF 443.00

CHF 249.00


Estimated to ship on14 April 2025



Select a Size

Change View
20 μG
CHF 249.00
50 μG
CHF 443.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

CHF 249.00


Estimated to ship on14 April 2025


description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

TTACTCGCCACAAGATGCTGCAGTTTGATGAGCTCTTCAAGCAGGAGACTGGCAAAGACAGGAAGATATCCAACACTTGGCTCAGCACTGGCTGGTTTACAATGACAATTGCACTGGAGCTCTGTGACAGGATCAATGTTTATGGCATGGTGCCCCCAGACTTCTGCAGGGATCCCAATCACCCTTCAGTACCTTATCATTATTATGAACCTTTTGGACCTGATGAATGTACAATGTACCTCTCCCATGAGCGAGGACGCAAGGGCAGTCATCACCGCTTTATCACAGAGAAACGAGTCTTTAAGAACTGGGCACGGACATTCAATATTCACTTTTTTCAACCAGACTGGAAACCAGAATCACTTGCTATAAATCATCCTGAGAATAAACCTGTGTTCTAAGGAATGAGCATGCCAGACTGTAATCCCAGGTATTCACTGCATCAGACACCGAGACACTGAACTTCCTGAGCCACCAGACAGGAAAGGGTAGCAGAAAACAGCTTCACTCCTCAGGAAGTACCATGGACAGACGCC

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Biochem/physiol Actions

ST6GALNAC5 (α-N-acetylgalactosaminide α-2,6-sialyltransferase 5) participates in the biosynthesis of α-series gangliosides. It is linked with breast cancer metastasis to the brain. ST6GALNAC5 reduces the association between breast cancer cells and human blood brain barrier.[1] Mutations in this gene are associated with coronary artery disease.[2]

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kolsoum InanlooRahatloo et al.
Scientific reports, 4, 3595-3595 (2014-01-09)
We aimed to identify the genetic cause of coronary artery disease (CAD) in an Iranian pedigree. Genetic linkage analysis identified three loci with an LOD score of 2.2. Twelve sequence variations identified by exome sequencing were tested for segregation with
ST6GALNAC5 Expression Decreases the Interactions between Breast Cancer Cells and the Human Blood-Brain Barrier.
Drolez A et al.
International Journal of Molecular Sciences, 17, E1309-E1309 (2016)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service