Skip to Content
Merck
All Photos(2)

Key Documents

E9783

Sigma-Aldrich

p3XFLAG-CMV-9 Expression Vector

Shuttle vector for transient or stable expression of secreted N-terminal 3xFLAG fusion proteins

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

tag

3X FLAG tagged

grade

for molecular biology

form

buffered aqueous solution

shipped in

dry ice

storage temp.

−20°C

General description

The p3XFLAG-CMV-9 Expression Vector is a 6.4 kb derivative of pCMV5 used to establish transient or stable expression of secreted N-terminal 3XFLAG fusion proteins in mammalian cells. The vector encodes three adjacent FLAG?epitopes (Asp-Tyr-Lys-Xaa-Xaa- Asp) upstream of the multiple cloning region. This results in increased detection sensitivity using ANTIFLAG M2 antibody. The third epitope includes the enterokinase recognition sequence, allowing cleavage of the 3XFLAG peptide from the purified fusion protein.

p3XFLAG-CMV-9 expression vector is a shuttle vector for E. coli and mammalian cells. Efficiency of replication and genomic integration is optimal when using an SV40 T antigen expressing host, such as COS cells. A related vector, p3XFLAG-CMV-3, has been used for stable transfection of HEK 293 cells. Efficiency of replication and genomic integration is optimal when using an SV40 T antigen-expressing host.

The p3XFLAG-CMV-7-BAP Control Plasmid is a 6.2 kb derivative of pCMV5 used for transient intracellular expression of N-terminal 3XFLAG bacterial alkaline phosphatase fusion protein in mammalian cells. The vector encodes three adjacent FLAG epitopes (Asp-Tyr-Lys-Xaa-Xaa-Asp) upstream of the multiple cloning region. This results in increased detection sensitivity using ANTI-FLAG M2 antibody. The third FLAG epitope includes the enterokinase recognition sequence, allowing cleavage of the 3XFLAG peptide from the purified fusion protein.

Vector Maps and Sequences

Components

  • p3XFLAG-CMV-9 Expression Vector 20 μg (E4276) is supplied as 0.5 mg/ml in 10 mM Tris-HCl (pH 8.0) with 1 mM EDTA.
  • p3XFLAG-CMV-7-BAP Control Plasmid 20 μg (C7472) is supplied as 0.5 mg/ml in 10 mM Tris-HCl (pH 8.0) with 1 mM EDTA.

Principle

The promoter-regulatory region of the human cytomegalovirus drives transcription of FLAG®-fusion constructs. The preprotrypsin leader sequence precedes the FLAG® sequence. The aminoglycoside phosphotransferase II gene (Neo-r) confers resistance to aminoglycosides such as G418 allowing for selection of stable transfectants.

Legal Information

This product is covered by the following patents owned by Sigma-Aldrich Co. LLC: US6,379,903, US7,094,548, JP4405125,EP1220933, CA2386471 and AU774216.
3xFLAG is a trademark of Sigma-Aldrich Co. LLC
FLAG is a registered trademark of Merck KGaA, Darmstadt, Germany
p3xFLAG-CMV is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sylvie Brassart-Pasco et al.
PloS one, 7(4), e29587-e29587 (2012-04-28)
NC1 domains from α1, α2, α3 and α6(IV) collagen chains were shown to exert anti-tumor or anti-angiogenic activities, whereas the NC1 domain of the α4(IV) chain did not show such activities so far. We demonstrate in the present paper that
Julia E Maxson et al.
The Journal of biological chemistry, 285(50), 39021-39028 (2010-10-13)
Hemojuvelin (HJV) is an important regulator of iron metabolism. Membrane-anchored HJV up-regulates expression of the iron regulatory hormone, hepcidin, through the bone morphogenic protein (BMP) signaling pathway by acting as a BMP co-receptor. HJV can be cleaved by the furin
Shuling Fan et al.
The Journal of cell biology, 178(3), 387-398 (2007-07-25)
The Crumbs family of apical transmembrane proteins regulates apicobasal polarity via protein interactions with a conserved C-terminal sequence, ERLI. However, one of the mammalian Crumbs proteins, Crumbs3 (CRB3) has an alternate splice form with a novel C-terminal sequence ending in
Shuntaro Oniki et al.
Cancer research, 66(12), 6395-6404 (2006-06-17)
Recent studies revealed that two novel interleukin (IL)-12-related cytokines, IL-23 and IL-27, have potent antitumor activities. However, the antitumor effects were mainly evaluated in relatively highly immunogenic tumors and have not been fully evaluated against nonimmunogenic or poorly immunogenic tumors.
Alexander David Barrow et al.
The Journal of clinical investigation, 121(9), 3505-3516 (2011-08-16)
Osteoclasts are terminally differentiated leukocytes that erode the mineralized bone matrix. Osteoclastogenesis requires costimulatory receptor signaling through adaptors containing immunoreceptor tyrosine-based activation motifs (ITAMs), such as Fc receptor common γ (FcRγ) and DNAX-activating protein of 12 kDa. Identification of these

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service