Skip to Content
Merck
All Photos(1)

Key Documents

E3285

Sigma-Aldrich

EIINFEKL trifluoroacetate salt

≥95% (HPLC)

Synonym(s):

Glu-Ile- Ile -Asn-Phe-Glu-Lys-Leu trifluoroacetate, L-a-glutamyl-L-isoleucyl-L-isoleucyl-L-asparaginyl-L-phenylalanyl-L-a-glutamyl-L-lysyl-L-Leucine trifluoroacetate, OVA – E1 Peptide

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
CHF 173.00
5 MG
CHF 556.00

CHF 173.00


Estimated to ship on29 May 2025



Select a Size

Change View
1 MG
CHF 173.00
5 MG
CHF 556.00

About This Item

Empirical Formula (Hill Notation):
C47H76N10O14 · xC2HF3O2
CAS Number:
Molecular Weight:
1005.16 (free base basis)
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

CHF 173.00


Estimated to ship on29 May 2025


Assay

≥95% (HPLC)

form

powder

color

white

solubility

acetonitrile: water: ~1 mg/mL (60/40)

storage temp.

−20°C

SMILES string

OC(=O)C(F)(F)F.CC[C@H](C)C(NC(=O)[C@@H](N)CCC(O)=O)C(=O)NC([C@@H](C)CC)C(=O)N[C@@H](CC(N)=O)C(=O)N[C@@H](Cc1ccccc1)C(=O)N[C@@H](CCC(O)=O)C(=O)N[C@@H](CCCCN)C(=O)N[C@@H](CC(C)C)C(O)=O

InChI

1S/C47H76N10O14.C2HF3O2/c1-7-26(5)38(57-46(69)39(27(6)8-2)56-40(63)29(49)17-19-36(59)60)45(68)54-33(24-35(50)58)44(67)53-32(23-28-14-10-9-11-15-28)43(66)52-31(18-20-37(61)62)42(65)51-30(16-12-13-21-48)41(64)55-34(47(70)71)22-25(3)4;3-2(4,5)1(6)7/h9-11,14-15,25-27,29-34,38-39H,7-8,12-13,16-24,48-49H2,1-6H3,(H2,50,58)(H,51,65)(H,52,66)(H,53,67)(H,54,68)(H,55,64)(H,56,63)(H,57,69)(H,59,60)(H,61,62)(H,70,71);(H,6,7)/t26-,27-,29-,30-,31-,32-,33-,34-,38-,39-;/m0./s1

InChI key

OAOVWFXRXDBOSX-YUPPFQQGSA-N

Amino Acid Sequence

Glu-Ile- Ile -Asn-Phe-Glu-Lys-Leu

Biochem/physiol Actions

EIINFEKL is an antagonist variant of SIINFEKL [OVA (257-264)] [it induces positive selection T-cells].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service