D9415
(2-Dodecen-1-yl)succinic anhydride
~95% (titration)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C16H26O3
CAS Number:
Molecular Weight:
266.38
EC Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
vapor density
9.2 (vs air)
vapor pressure
<1 mmHg ( 20 °C)
Assay
~95% (titration)
bp
180-182 °C/5 mmHg (lit.)
mp
41-43 °C (lit.)
SMILES string
CCCCCCCCC\C=C/CC1CC(=O)OC1=O
InChI
1S/C16H26O3/c1-2-3-4-5-6-7-8-9-10-11-12-14-13-15(17)19-16(14)18/h10-11,14H,2-9,12-13H2,1H3/b11-10+
InChI key
UYCICMIUKYEYEU-ZHACJKMWSA-N
Looking for similar products? Visit Product Comparison Guide
replaced by
Product No.
Description
Pricing
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
352.4 °F - closed cup
Flash Point(C)
178 °C - closed cup
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Vinod Vellora Thekkae Padil et al.
Journal of agricultural and food chemistry, 63(14), 3757-3765 (2015-03-24)
Esterifications of the tree-based gum, gum karaya (GK), using dodecenylsuccinic anhydride (DDSA) were carried out in aqueous solutions. GK was deacetylated using alkali treatment to obtain deacetylated gum karaya (DGK). The DGK and its DDSA derivative were characterized using gel
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service