Skip to Content
Merck
All Photos(1)

Key Documents

D5511

Sigma-Aldrich

N,N-Dimethylacetamide

≥99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CON(CH3)2
CAS Number:
Molecular Weight:
87.12
Beilstein:
1737614
EC Number:
MDL number:
UNSPSC Code:
12352111
PubChem Substance ID:

vapor density

3 (vs air)

vapor pressure

2 mmHg ( 25 °C)
4 mmHg ( 38 °C)

Assay

≥99%

autoignition temp.

914 °F

expl. lim.

1.8 %, 100 °F
11.5 %, 160 °F

refractive index

n20/D 1.437 (lit.)

bp

164.5-166 °C (lit.)

mp

−20 °C (lit.)

density

0.937 g/mL at 25 °C (lit.)

SMILES string

CN(C)C(C)=O

InChI

1S/C4H9NO/c1-4(6)5(2)3/h1-3H3

InChI key

FXHOOIRPVKKKFG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Eye Irrit. 2 - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

157.1 °F - closed cup

Flash Point(C)

69.5 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yanli Yang et al.
mAbs, 11(6), 996-1011 (2019-06-04)
Agonism of members of the tumor necrosis factor receptor superfamily (TNFRSF) with monoclonal antibodies is of high therapeutic interest due to their role in immune regulation and cell proliferation. A major hurdle for pharmacologic activation of this receptor class is
Inês Farinha et al.
Carbohydrate polymers, 130, 455-464 (2015-06-17)
Purified chitin-glucan complex (CGCpure) was extracted from Komagataella pastoris biomass using a hot alkaline treatment, followed by neutralization and repeated washing with deionized water. The co-polymer thus obtained had a β-glucan:chitin molar ratio of 75:25 and low protein and inorganic
S Sood et al.
Animal reproduction science, 129(1-2), 89-95 (2011-11-15)
Three experiments conducted to improve the survival of emu sperm during cryopreservation aimed to: (1) minimize chilling injury during the cooling phase; (2) determine the osmotic effects of dimethylacetamide (DMA), sucrose and trehalose; and (3) investigate the timing and nature
Tony Reid et al.
The Lancet. Oncology, 16(9), 1133-1142 (2015-08-25)
Epigenetic alterations have been strongly associated with tumour formation and resistance to chemotherapeutic drugs, and epigenetic modifications are an attractive target in cancer research. RRx-001 is activated by hypoxia and induces the generation of reactive oxygen and nitrogen species that
Jingshuai Yang et al.
ChemSusChem, 6(2), 275-282 (2013-01-11)
Covalently cross-linked polymer membranes were fabricated from poly(aryl sulfone benzimidazole) (SO(2)PBI) and poly(vinylbenzyl chloride) (PVBCl) as electrolytes for high-temperature proton-exchange-membrane fuel cells. The cross-linking imparted organo insolubility and chemical stability against radical attack to the otherwise flexible SO(2)PBI membranes. Steady

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service