Select a Size
All Photos(1)
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C6H12Br2O4
CAS Number:
Molecular Weight:
307.97
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
storage temp.
−20°C
SMILES string
O[C@H](CBr)[C@@H](O)[C@H](O)[C@H](O)CBr
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service