All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C10H16N2O5
CAS Number:
Molecular Weight:
244.24
MDL number:
UNSPSC Code:
12352204
storage temp.
−20°C
SMILES string
CC1CN(C2CC(O)C(CO)O2)C(=O)NC1=O
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
M A Chaudhry et al.
Journal of molecular biology, 249(5), 914-922 (1995-06-23)
Energy deposition by ionizing radiation can lead to the formation of clustered DNA damage, i.e. more than one lesion situated within a helical turn of DNA. Among the postulated lesions are those characterized by damaged bases and abasic sites on
Efficiency of incision of an AP site within clustered DNA damage by the major human AP endonuclease.
Marie-Hélène David-Cordonnier et al.
Biochemistry, 41(2), 634-642 (2002-01-10)
A major DNA lesion induced by ionizing radiation and formed on removal of oxidized base lesions by various glycosylases is an apurinic/apyrimidinic site (AP site). The presence of an AP site within clustered DNA damage, induced following exposure to ionizing
E A Furlong et al.
Biochemistry, 25(15), 4344-4349 (1986-07-29)
5,6-Dihydrothymidine (dDHT) is a derivative thymidine formed during gamma-irradiation. This paper demonstrates the conditions under which dDHT is formed in solutions of DNA and that dDHT is produced in the DNA of HeLa cells during gamma-irradiation. The product of dDHT
M Weinfeld et al.
Nucleic acids research, 21(3), 621-626 (1993-02-11)
Stacking between aromatic amino acids and nucleic acid bases may play an important role in the interaction of enzymes with nucleic acid substrates. In such circumstances, disruption of base aromaticity would be expected to decrease enzyme activity on the modified
J Cadet et al.
International journal of radiation biology and related studies in physics, chemistry, and medicine, 39(2), 119-133 (1981-02-01)
The steady-state gamma radiolysis of deaerated aqueous solutions of thymidine generated a complex mixture of pyrimidine and nucleoside derivatives. Twenty-two of these compounds have been isolated and unambiguously characterized by spectroscopic methods including proton nuclear magnetic resonance and mass spectrometry.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service